Merck
  • Application of CE-MS for the analysis of histones and histone modifications.
Application of CE-MS for the analysis of histones and histone modifications.
Methods (San Diego, Calif.) (2020-02-06)
Klaus Faserl, Bettina Sarg, Herbert H Lindner
ABSTRACT
The analysis, identification and quantification of histones and their post-translational modifications plays a central role in chromatin research and in studying epigenetic regulations during physiological processes. In the last decade analytical strategies based on mass spectrometry have been greatly improved for providing a global view of single modification abundances or to determine combinatorial patterns of modifications. Presented here is a newly developed strategy for histone protein analysis and a number of applications are illustrated with an emphasis on PTM characterization. Capillary electrophoresis is coupled to mass spectrometry (CE-MS) and has proven to be a very promising concept as it enables to study intact histones (top-down proteomics) as well as the analysis of enzymatically digested proteins (bottom-up proteomics). This technology combines highly efficient low-flow CE separations with ionization in a single device and offers an orthogonal separation principle to conventional LC-MS analysis, thus expanding the existing analytical repertoire in a perfect manner.
Product Description
Supelco
Acetic acid 100%, for LC-MS LiChropur
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Pricing and availability is not currently available.