MilliporeSigma
  • Blue Light Mediates Chloroplast Avoidance and Enhances Photoprotection of Vanilla Orchid.
Blue Light Mediates Chloroplast Avoidance and Enhances Photoprotection of Vanilla Orchid.
International journal of molecular sciences (2020-11-01)
Swee-Suak Ko, Chung-Min Jhong, Yi-Jyun Lin, Ching-Yu Wei, Ju-Yin Lee, Ming-Che Shih
ABSTRACT
Vanilla orchid, which is well-known for its flavor and fragrance, is cultivated in tropical and subtropical regions. This shade-loving plant is very sensitive to high irradiance. In this study, we show that vanilla chloroplasts started to have avoidance movement when blue light (BL) was higher than 20 μmol m-2s-1 and significant avoidance movement was observed under BL irradiation at 100 μmol m-2s-1 (BL100). The light response curve indicated that when vanilla was exposed to 1000 μmol m-2s-1, the electron transport rate (ETR) and photochemical quenching of fluorescence (qP) were significantly reduced to a negligible amount. We found that if a vanilla orchid was irradiated with BL100 for 12 days, it acquired BL-acclimation. Chloroplasts moved to the side of cells in order to reduce light-harvesting antenna size, and chloroplast photodamage was eliminated. Therefore, BL-acclimation enhanced vanilla orchid growth and tolerance to moderate (500 μmol m-2s-1) and high light (1000 μmol m-2s-1) stress conditions. It was found that under high irradiation, BL-acclimatized vanilla maintained higher ETR and qP capacity than the control without BL-acclimation. BL-acclimation induced antioxidant enzyme activities, reduced ROS accumulation, and accumulated more carbohydrates. Moreover, BL-acclimatized orchids upregulated photosystem-II-associated marker genes (D1 and PetC), Rubisco and PEPC transcripts and sustained expression levels thereof, and also maximized the photosynthesis rate. Consequently, BL-acclimatized orchids had higher biomass. In short, this study found that acclimating vanilla orchid with BL before transplantation to the field might eliminate photoinhibition and enhance vanilla growth and production.