Merck
T Cell Activation Depends on Extracellular Alanine.
Cell reports (2019-09-19)
Noga Ron-Harel, Jonathan M Ghergurovich, Giulia Notarangelo, Martin W LaFleur, Yoshiki Tsubosaka, Arlene H Sharpe, Joshua D Rabinowitz, Marcia C Haigis
ABSTRACT
T cell stimulation is metabolically demanding. To exit quiescence, T cells rely on environmental nutrients, including glucose and the amino acids glutamine, leucine, serine, and arginine. The expression of transporters for these nutrients is tightly regulated and required for T cell activation. In contrast to these amino acids, which are essential or require multi-step biosynthesis, alanine can be made from pyruvate by a single transamination. Here, we show that extracellular alanine is nevertheless required for efficient exit from quiescence during naive T cell activation and memory T cell restimulation. Alanine deprivation leads to metabolic and functional impairments. Mechanistically, this vulnerability reflects the low expression of alanine aminotransferase, the enzyme required for interconverting pyruvate and alanine, whereas activated T cells instead induce alanine transporters. Stable isotope tracing reveals that alanine is not catabolized but instead supports protein synthesis. Thus, T cells depend on exogenous alanine for protein synthesis and normal activation.
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
Anti-Puromycin, clone 12D10, Alexa Fluor 647 Conjugate Antibody, clone 12D10, 0.5 mg/mL, from mouse
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder