Merck
  • Induction of UCP1 and thermogenesis by a small molecule via AKAP1/PKA modulation.
Induction of UCP1 and thermogenesis by a small molecule via AKAP1/PKA modulation.
The Journal of biological chemistry (2020-08-29)
Laurent Vergnes, Jason Y Lin, Graeme R Davies, Christopher D Church, Karen Reue, Laurent Vergnes, Jason Y Lin, Graeme R Davies, Christopher D Church, Karen Reue
ABSTRACT
Strategies to increase energy expenditure are an attractive approach to reduce excess fat storage and body weight to improve metabolic health. In mammals, uncoupling protein-1 (UCP1) in brown and beige adipocytes uncouples fatty acid oxidation from ATP generation in mitochondria and promotes energy dissipation as heat. We set out to identify small molecules that enhance UCP1 levels and activity using a high-throughput screen of nearly 12,000 compounds in mouse brown adipocytes. We identified a family of compounds that increase Ucp1 expression and mitochondrial activity (including un-coupled respiration) in mouse brown adipocytes and human brown and white adipocytes. The mechanism of action may be through compound binding to A kinase anchoring protein (AKAP) 1, modulating its localization to mitochondria and its interaction with protein kinase A (PKA), a known node in the β-adrenergic signaling pathway. In mice, the hit compound increased body temperature, UCP1 protein levels, and thermogenic gene expression. Some of the compound effects on mitochondrial function were UCP1- or AKAP1-independent, suggesting compound effects on multiple nodes of energy regulation. Overall, our results highlight a role for AKAP1 in thermogenesis, uncoupled respiration, and regulation energy balance.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
AST Activity Assay Kit, sufficient for 100 colorimetric tests
Sigma-Aldrich
Anti-PGC-1 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Uncoupling Protein-1 (UCP-1) (145-159) Rabbit pAb, liquid, Calbiochem®