Merck
  • Activity of B-nor analogues of neurosteroids on the GABA(A) receptor in primary neuronal cultures.
Activity of B-nor analogues of neurosteroids on the GABA(A) receptor in primary neuronal cultures.
Journal of medicinal chemistry (2006-05-26)
Cristina Suñol, Daniel A García, Jordi Bujons, Zdena Kristofíková, Libor Matyás, Zoila Babot, Alexander Kasal
ABSTRACT
A GABA(A) receptor study of several B-nor analogues of allopregnanolone and pregnanolone has been carried out. B-norallopregnanolone (i.e., 3alpha-hydroxy-7-nor-5alpha-pregnan-20-one) was found comparable to allopregnanolone when measured with labeled TBPS. Analogous results were obtained from their effect on neurons in culture: this time, both 3alpha-hydroxy-7-nor-5xi-pregnan-20-ones (5 and 6) were found to stimulate [3H]flunitrazepam binding and GABA-induced 36Cl- influx. These effects were inhibited by GABA(A) receptor antagonists. Other analogues carrying electronegative substituents (epoxides 9 and 10 and ketone 12) in the B ring were inactive. Similarly, B-normal ketones 17, and 18 and 6-azasteroids 20 and 21 were also inactive. B-Nor analogues 5 and 6 did not induce neurotoxicity at relevant concentrations. A computational analysis of active and inactive neurosteroid analogues allowed the proposal of a 3D pharmacophoric hypothesis of their interaction with the GABA(A) receptor.