Merck
  • Functional analyses of mammalian virus 5'UTR-derived, small RNAs that regulate virus translation.
Functional analyses of mammalian virus 5'UTR-derived, small RNAs that regulate virus translation.
Methods (San Diego, Calif.) (2020-02-23)
Mei-Ling Li, Gary Brewer, Mei-Ling Li, Gary Brewer
ABSTRACT
Enterovirus A71 (EV-A711) RNA contains an internal ribosomal entry site (IRES) to direct cap-independent translation. IRES-dependent translation requires the host's translation initiation factors and IRES-associated trans-acting factors (ITAFs). We previously showed that hnRNP A1, the mRNA stability factor HuR, and the RISC subunit Argonaute 2 (Ago2) are ITAFs that associate with stem loop II (SL-II) of the IRES and promote IRES-dependent translation. By contrast, the mRNA decay factor AUF1 is a negative-acting ITAF that also binds SL-II. Moreover, the small RNA-processing enzyme Dicer produces at least four virus-derived, small RNAs (vsRNAs 1-4) from the EV-A71 5'UTR in infected cells. One of these, vsRNA1, derived from SL-II, inhibits IRES activity via an unknown mechanism. In vitro RNA-binding assays revealed that vsRNA1 can alter association of Ago2, HuR, and AUF1 with SL-II. This presents a possible mechanism by which vsRNA1 could control association of ITAFs with the IRES and modulate viral translation. Here, we describe methods for functional analyses of vsRNA1-mediated regulation of IRES activity. These methods should be applicable to other virus-derived, small RNAs as well.
Sigma-Aldrich
MISSION® esiRNA, targeting human HNRNPA1
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution