Merck
  • Epicardial cell shape and maturation are regulated by Wt1 via transcriptional control of Bmp4.
Epicardial cell shape and maturation are regulated by Wt1 via transcriptional control of Bmp4.
Development (Cambridge, England) (2019-10-19)
Víctor Velecela, Alejo Torres-Cano, Ana García-Melero, Marina Ramiro-Pareta, Claudia Müller-Sánchez, Marc Segarra-Mondejar, You-Ying Chau, Begoña Campos-Bonilla, Manuel Reina, Francesc X Soriano, Nicholas D Hastie, Fernando O Martínez, Ofelia M Martínez-Estrada
ABSTRACT
The epicardium plays a crucial role in embryonic heart development and adult heart repair; however, the molecular events underlying its maturation remain unknown. Wt1, one of the main markers of the embryonic epicardium, is essential for epicardial development and function. Here, we analyse the transcriptomic profile of epicardial-enriched cells at different stages of development and from control and epicardial-specific Wt1 knockout (Wt1KO) mice. Transcriptomic and cell morphology analyses of epicardial cells from epicardial-specific Wt1KO mice revealed a defect in the maturation process of the mutant epicardium, including sustained upregulation of Bmp4 expression and the inability of mutant epicardial cells to transition into a mature squamous phenotype. We identified Bmp4 as a transcriptional target of Wt1, thus providing a molecular basis for the retention of the cuboidal cell shape observed in the Wt1KO epicardium. Accordingly, inhibition of the Bmp4 signalling pathway both ex vivo and in vivo rescued the cuboidal phenotype of the mutant epicardium. Our findings indicate the importance of the cuboidal-to-squamous transition in epicardial maturation, a process regulated by Wt1.
Product Description
Roche
DIG RNA Labeling Kit (SP6/T7), sufficient for 2 x 10 labeling reactions, kit of 1 (12 components), suitable for hybridization, suitable for Southern blotting
Sigma-Aldrich
IgG from rabbit serum, technical grade, ≥80% (SDS-PAGE), buffered aqueous solution