Merck
  • A diarylquinoline drug active on the ATP synthase of Mycobacterium tuberculosis.
A diarylquinoline drug active on the ATP synthase of Mycobacterium tuberculosis.
Science (New York, N.Y.) (2004-12-14)
Koen Andries, Peter Verhasselt, Jerome Guillemont, Hinrich W H Göhlmann, Jean-Marc Neefs, Hans Winkler, Jef Van Gestel, Philip Timmerman, Min Zhu, Ennis Lee, Peter Williams, Didier de Chaffoy, Emma Huitric, Sven Hoffner, Emmanuelle Cambau, Chantal Truffot-Pernot, Nacer Lounis, Vincent Jarlier
ABSTRACT
The incidence of tuberculosis has been increasing substantially on a worldwide basis over the past decade, but no tuberculosis-specific drugs have been discovered in 40 years. We identified a diarylquinoline, R207910, that potently inhibits both drug-sensitive and drug-resistant Mycobacterium tuberculosis in vitro (minimum inhibitory concentration 0.06 mug/ml). In mice, R207910 exceeded the bactericidal activities of isoniazid and rifampin by at least 1 log unit. Substitution of drugs included in the World Health Organization's first-line tuberculosis treatment regimen (rifampin, isoniazid, and pyrazinamide) with R207910 accelerated bactericidal activity, leading to complete culture conversion after 2 months of treatment in some combinations. A single dose of R207910 inhibited mycobacterial growth for 1 week. Plasma levels associated with efficacy in mice were well tolerated in healthy human volunteers. Mutants selected in vitro suggest that the drug targets the proton pump of adenosine triphosphate (ATP) synthase.
Product Description
Supelco
Isoniazid, analytical standard, ≥99% (TLC)
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
TMC207, ≥98% (HPLC)