Merck
  • C1 Domain-targeted isophthalate derivatives induce cell elongation and cell cycle arrest in HeLa cells.
C1 Domain-targeted isophthalate derivatives induce cell elongation and cell cycle arrest in HeLa cells.
PloS one (2011-06-02)
Virpi Talman, Raimo K Tuominen, Gustav Boije af Gennäs, Gustav Boije Af Gennäs, Jari Yli-Kauhaluoma, Elina Ekokoski
ABSTRACT
Diacylglycerol (DAG)-mediated signaling pathways, such as those mediated by protein kinase C (PKC), are central in regulating cell proliferation and apoptosis. DAG-responsive C1 domains are therefore considered attractive drug targets. Our group has designed a novel class of compounds targeted to the DAG binding site within the C1 domain of PKC. We have previously shown that these 5-(hydroxymethyl)isophthalates modulate PKC activation in living cells. In this study we investigated their effects on HeLa human cervical cancer cell viability and proliferation by using standard cytotoxicity tests and an automated imaging platform with machine vision technology. Cellular effects and their mechanisms were further characterized with the most potent compound, HMI-1a3. Isophthalate derivatives with high affinity to the PKC C1 domain exhibited antiproliferative and non-necrotic cytotoxic effects on HeLa cells. The anti-proliferative effect was irreversible and accompanied by cell elongation. HMI-1a3 induced down-regulation of retinoblastoma protein and cyclins A, B1, D1, and E. Effects of isophthalates on cell morphology, cell proliferation and expression of cell cycle-related proteins were different from those induced by phorbol 12-myristate-13-acetate (PMA) or bryostatin 1, but correlated closely to binding affinities. Therefore, the results strongly indicate that the effect is C1 domain-mediated.
Sigma-Aldrich
Isophthalic acid, 99%
Sigma-Aldrich
Boc-Asp(OMe)-fluoromethyl ketone, ≥90% (TLC), solid
Sigma-Aldrich
Bryostatin 1, ≥99%, solid