Merck
An alternative and simple synthesis of [14 C]potassium cyanate.
Journal of labelled compounds & radiopharmaceuticals (2020-02-15)
Crist N Filer, C T Peng
ABSTRACT
The one-step synthesis of [14 C]potassium cyanate from [14 C]urea is described with product characterization by gravimetric specific activity as well as a novel TLC system. The storage, stability, and repurification of [14 C]potassium cyanate are also discussed.
Sigma-Aldrich
Potassium cyanate, 96%
Sigma-Aldrich
4-Methyl-2-pentanol, 98%