Merck
  • Dehydroabietic acid alleviates high fat diet-induced insulin resistance and hepatic steatosis through dual activation of PPAR-γ and PPAR-α.
Dehydroabietic acid alleviates high fat diet-induced insulin resistance and hepatic steatosis through dual activation of PPAR-γ and PPAR-α.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2020-05-16)
Zhishen Xie, Gai Gao, Hui Wang, Erwen Li, Yong Yuan, Jiangyan Xu, Zhenqiang Zhang, Pan Wang, Yu Fu, Huahui Zeng, Junying Song, Christian Hölscher, Hui Chen
ABSTRACT
Dual-PPAR-α/γ agonist has the dual potentials to improve insulin resistance (IR) and hepatic steatosis associated with obesity. This study aimed to investigate whether dehydroabietic acid (DA), a naturally occurred compound, can bind to and activate both PPAR-γ and PPAR-α to ameliorate IR and hepatic steatosis in high-fat diet (HFD)-fed mice.. We found that DA formed stable hydrogen bonds with the ligand-binding domains of PPAR-γ and PPAR-α. DA treatment also promoted 3T3-L1 differentiation via PPAR-γ activation, and mitochondrial oxygen consumption in HL7702 cells via PPAR-α activation. In HFD-fed mice, DA treatment alleviated glucose intolerance and IR, and reduced hepatic steatosis, liver injury markers (ALT, AST), and lipid accumulation, and promoted mRNA expression of PPAR-γ and PPAR-α signaling elements involved in IR and lipid metabolism in vivo and in vitro, and inhibited mRNA expression of pro-inflammatory factors. Therefore, DA is a dual-PPAR-α/γ and PPAR-γ partial agonist, which can attenuate IR and hepatic steatosis induced by HFD-consumption in mice.
Sigma-Aldrich
Abietic acid, technical, ~75% (GC)