Merck
  • Intracellular Role for the Matrix-Modifying Enzyme Lox in Regulating Transcription Factor Subcellular Localization and Activity in Muscle Regeneration.
Intracellular Role for the Matrix-Modifying Enzyme Lox in Regulating Transcription Factor Subcellular Localization and Activity in Muscle Regeneration.
Developmental cell (2020-05-04)
Ravit Gabay Yehezkely, Shelly Zaffryar-Eilot, Anna Kaganovsky, Nurit Fainshtain Malka, Rohtem Aviram, Ido Livneh, Peleg Hasson
ABSTRACT
Integration of extracellular matrix (ECM)-derived cues into transcriptional programs is essential primarily in rapidly morphing environments, such as regenerating tissues. Here, we demonstrate that lysyl oxidase (Lox), known for its ECM-modifying activities, primarily collagen crosslinking, also directly regulates transcription factor (TF) localization. Using genetic and pharmacological strategies, we highlight an intracellular role for Lox in myogenic progenitors essential for muscle regeneration. We show that Lox interacts with, and directly oxidizes, vestigial-like 3 (Vgll3), a transcriptional co-activator acting with Mef2 and transcriptional enhancer factor (TEF) TFs. This enzymatic activity is required for Vgll3 cytoplasmic-to-nuclear translocation in regulation of myogenic differentiation. Our work highlights an additional mechanism for TF subcellular localization facilitating integration of ECM organization with transcriptional output during myogenic differentiation. Modulating this integration mechanism could affect the balance between ECM organization and cell differentiation and serve as a basis for novel therapeutic strategies targeting fibrotic pathologies.
Sigma-Aldrich
Anti-NOL3 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
MISSION® pLKO.1-puro Non-Target shRNA Control Plasmid DNA, Targets no known genes from any species
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
3-Aminopropionitrile fumarate salt, metabolite