Merck
  • Functional Evaluation of AMD-Associated Risk Variants of Complement Factor B.
Functional Evaluation of AMD-Associated Risk Variants of Complement Factor B.
Investigative ophthalmology & visual science (2020-05-15)
Camilla Pilotti, John Greenwood, Stephen E Moss
ABSTRACT
The 32W and 32Q variants of complement factor B (CFB) are associated with reduced risk of developing neovascular age-related macular degeneration (AMD) compared with the common 32R allele. The objective of this study was to determine if the most protective R32Q variant affects the neovascular process in a manner consistent with the reported reduced disease association. The 32R, 32W, and 32Q human CFB variants were expressed in human embryonic kidney 293T cells and purified from culture supernatant. The ex vivo mouse fetal metatarsal explant model was used to investigate the effect of these three human CFB variants on angiogenesis. Metatarsal bones were isolated from mouse embryos and cultured in the presence of the three CFB variants, and angiogenesis was measured following immunostaining of fixed samples. ELISAs were used to quantify C3 and VEGF protein levels in metatarsal culture and quantitative PCR to measure Cfb, C3, and Vegf expression. We show here that the three CFB variants have different biological activities in the mouse metatarsal assay, with CFBR32 exhibiting significantly greater angiogenic activity than CFBQ32 or CFBW32, which were broadly similar. We also observed differences in macrophage phenotype with these two variants that may contribute to their activities in this experimental model. We have demonstrated that the biological activities of CFBR32, CFBW32, and CFBQ32 are consistent with their AMD risk association, and we provide functional evidence of roles for these variants in angiogenesis that may be relevant to the pathogenesis of the neovascular form of AMD.
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
Brilliant Blue R, 250, suitable for microscopy