Merck
Autophagy as a decisive process for cell death.
Experimental & molecular medicine (2020-06-28)
Seonghee Jung, Hyeonjeong Jeong, Seong-Woon Yu
ABSTRACT
Autophagy is an intracellular catabolic pathway in which cellular constituents are engulfed by autophagosomes and degraded upon autophagosome fusion with lysosomes. Autophagy serves as a major cytoprotective process by maintaining cellular homeostasis and recycling cytoplasmic contents. However, emerging evidence suggests that autophagy is a primary mechanism of cell death (autophagic cell death, ACD) and implicates ACD in several aspects of mammalian physiology, including tumor suppression and psychological disorders. However, little is known about the physiological roles and molecular mechanisms of ACD. In this review, we document examples of ACD and discuss recent progress in our understanding of its molecular mechanisms.
Sigma-Aldrich
Neferine, ≥98% (HPLC)