Merck
  • Mercury and Organochlorine Pesticides in Tissues of Loggerhead Sea Turtles (Caretta caretta) Stranded Along the Southwestern Mediterranean Coastline (Andalusia, Spain).
Mercury and Organochlorine Pesticides in Tissues of Loggerhead Sea Turtles (Caretta caretta) Stranded Along the Southwestern Mediterranean Coastline (Andalusia, Spain).
Bulletin of environmental contamination and toxicology (2020-03-19)
Pilar Gómez-Ramírez, Silvia Espín, Isabel Navas, Emma Martínez-López, Pedro Jiménez, Pedro María-Mojica, José Peñalver, Antonio J García-Fernández
ABSTRACT
Nineteen loggerhead sea turtles (Caretta caretta) stranded along the southwestern Mediterranean coastline (Andalusia) were used in this study. A total of 68 samples of fat (n = 18), liver (n = 15), kidney (n = 13), pectoral muscle (n = 19), and brain (n = 3) were analysed for total mercury (Hg) and organochlorine pesticides [OC: ∑Dichlorodiphenyltrichloroethanes (∑DDT), ∑Hexachlorocyclohexane (∑HCH), ∑Heptachlor, ∑Drins and ∑Endosulfan]. These loggerhead sea turtles showed tissue Hg and OC concentrations similar to or lower than those reported in other studies. Few growth-related variations in Hg or OC levels in relation to straight carapace length were found, probably because the specimens were mostly juveniles. This study will help to fill the gap on spatio-temporal exposure data and ascertain the real world-wide picture of the contamination levels in loggerhead sea turtles.
Product Description
Supelco
EPA Pesticide Mix, in methanol: methylene chloride (98:2) (varied), TraceCERT®, analytical standard
Supelco
Endosulfan, PESTANAL®, analytical standard