Merck
  • Distinct calcium/calmodulin-dependent serine protein kinase domains control cardiac sodium channel membrane expression and focal adhesion anchoring.
Distinct calcium/calmodulin-dependent serine protein kinase domains control cardiac sodium channel membrane expression and focal adhesion anchoring.
Heart rhythm (2020-01-07)
Adeline Beuriot, Catherine A Eichel, Gilles Dilanian, Florent Louault, Dario Melgari, Nicolas Doisne, Alain Coulombe, Stéphane N Hatem, Elise Balse
ABSTRACT
Membrane-associated guanylate kinase proteins function as adaptor proteins to mediate the recruitment and scaffolding of ion channels in the plasma membrane in various cell types. In the heart, the protein calcium/calmodulin-dependent serine protein kinase (CASK) negatively regulates the main cardiac sodium channel NaV1.5, which carries the sodium current (INa) by preventing its anterograde trafficking. CASK is also a new member of the dystrophin-glycoprotein complex and, like syntrophin, binds to the C-terminal domain of the channel. The purpose of this study was to unravel the mechanisms of CASK-mediated negative INa regulation and interaction with the dystrophin-glycoprotein complex in cardiac myocytes. CASK adenoviral truncated constructs with sequential single functional domain deletions were designed for overexpression in cardiac myocytes: CASKΔCAMKII, CASKΔL27A, CASKΔL27B, CASKΔPDZ, CASKΔSH3, CASKΔHOOK, and CASKΔGUK. A combination of whole-cell patch-clamp recording, total internal reflection fluorescence microscopy, and biochemistry experiments was conducted in cardiac myocytes to study the functional consequences of domain deletions. We show that both L27B and GUK domains are required for the negative regulatory effect of CASK on INa and NaV1.5 surface expression and that the HOOK domain is essential for interaction with the cell adhesion dystrophin-glycoprotein complex. This study demonstrates that the multimodular structure of CASK confers an ability to simultaneously interact with several targets within cardiomyocytes. Through its L27B, GUK, and HOOK domains, CASK potentially provides the ability to control channel delivery at adhesion points in cardiomyocytes.
Sigma-Aldrich
Monoclonal Anti-Dystrophin antibody produced in mouse, clone MANDYS8, ascites fluid
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Anti-CASK Antibody, clone 2G5.1, clone 2G5.1, from mouse