Merck
Injury Delays Stem Cell Apoptosis after Radiation in Planarians.
Current biology : CB (2020-05-11)
Divya A Shiroor, Tisha E Bohr, Carolyn E Adler
ABSTRACT
Stem cells are continuously exposed to multiple stresses, including radiation and tissue injury. As central drivers of tissue repair and regeneration, it is necessary to understand how their behavior is influenced by these stressors. Planarians have an abundant population of stem cells that are rapidly eliminated after radiation exposure via apoptosis. Low doses of radiation eliminate the majority of these stem cells, allowing a few to remain [1]. Here, we combine radiation with injury to define how stem cells respond to tissue damage. We find that a variety of injuries induced within a defined window of time surrounding radiation cause stem cells to outlast those in uninjured animals. Injury stimulates localized cell death adjacent to wounds [2], in the same regions where stem cells persist. This persistence occurs in the absence of proliferation. Instead, stem cells are retained near the wound due to delayed apoptosis, which we quantify by combining fluorescence-activated cell sorting (FACS) with annexin V staining. Pharmacological inhibition of the mitogen-activated protein (MAP) kinase extracellular signal-regulated kinase (ERK) prevents stem cell persistence after injury, implicating wound-induced ERK activity in this response. By combining radiation with injury, our work reveals a novel connection between dying cells and stem cells that remain. Furthermore, the ability to induce stem cell persistence after radiation provides a paradigm to study mechanisms that may contribute to unanticipated consequences of injury, such as tumorigenesis.
Sigma-Aldrich
ApopTag TdT Enzyme, ApopTag Terminal deoxynucleotidyl Transferase (TdT) is a recombinant enzyme intended for use in labeling the 3′-OH ends of fragmented DNA during apoptotic cell detection.
Roche
Anti-Digoxigenin-POD, Fab fragments, from sheep
Roche
Anti-Fluorescein-POD, Fab fragments, from sheep
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep