Merck
  • The DNA topoisomerase II inhibitor amsacrine as a novel candidate adjuvant in a model of glaucoma filtration surgery.
The DNA topoisomerase II inhibitor amsacrine as a novel candidate adjuvant in a model of glaucoma filtration surgery.
Scientific reports (2019-12-19)
Kotaro Yamamoto, Taiki Kokubun, Kota Sato, Takahiro Akaishi, Atsushi Shimazaki, Masatsugu Nakamura, Yukihiro Shiga, Satoru Tsuda, Kazuko Omodaka, Hideyuki Saya, Toru Nakazawa
ABSTRACT
Treatments for refractory glaucoma include trabeculectomy, in which a filtering bleb is created to reduce aqueous pressure. Mitomycin C (MMC) is often used as an adjuvant to reduce post-trabeculectomy bleb scarring and consequent failure. However, scarring sometimes still occurs. Thus, we searched for more effective trabeculectomy adjuvants with high-throughput screening (HTS) of a library of 1,165 off-patent drug compounds. This revealed that amsacrine (AMSA), a DNA topoisomerase II (TOP2) inhibitor, was the top candidate. Compared to MMC, rabbits that underwent trabeculectomy with 10% AMSA had lower IOP at 42, 56, and 70 days (P < 0.01 at all measurement points) and a higher bleb score at 28, 42, 56, and 70 days (P =  < 0.01, 0.04, 0.04, and < 0.01, respectively). Compared to saline, rabbits that received 1% AMSA also had lower IOP and better bleb score at all time points, without a sharp drop in IOP just after surgery (all P < 0.01). Both effects were milder than MMC at 7 days (P = 0.02 and <0.01, respectively). Thus, this study showed that HTS may help identify new, promising uses for off-patent drugs. Furthermore, trabeculectomy with AMSA at a suitable concentration may improve the prognosis after trabeculectomy compared to MMC.
Sigma-Aldrich
Amsacrine hydrochloride, ≥98% (TLC), powder