Merck
  • Antiviral activity of the dihydropyrone PNU-140690, a new nonpeptidic human immunodeficiency virus protease inhibitor.
Antiviral activity of the dihydropyrone PNU-140690, a new nonpeptidic human immunodeficiency virus protease inhibitor.
Antimicrobial agents and chemotherapy (1997-05-01)
S M Poppe, D E Slade, K T Chong, R R Hinshaw, P J Pagano, M Markowitz, D D Ho, H Mo, R R Gorman, T J Dueweke, S Thaisrivongs, W G Tarpley
ABSTRACT
PNU-140690 is a member of a new class of nonpeptidic human immunodeficiency virus (HIV) protease inhibitors (sulfonamide-containing 5,6-dihydro-4-hydroxy-2-pyrones) discovered by structure-based design. PNU-140690 has excellent potency against a variety of HIV type 1 (HIV-1) laboratory strains and clinical isolates, including those resistant to the reverse transcriptase inhibitors zidovudine or delavirdine. When combined with either zidovudine or delavirdine, PNU-140690 contributes to synergistic antiviral activity. PNU-140690 is also highly active against HIV-1 variants resistant to peptidomimetic protease inhibitors, underscoring the structural distinctions between PNU-140690 and substrate analog protease inhibitors. PNU-140690 retains good antiviral activity in vitro in the presence of human plasma proteins, and preclinical pharmacokinetic studies revealed good oral bioavailability. Accordingly, PNU-140690 is a candidate for clinical evaluation.
Sigma-Aldrich
Tipranavir, ≥98% (HPLC)