Merck
  • Circular RNA hsa_circ_0061825 (circ-TFF1) contributes to breast cancer progression through targeting miR-326/TFF1 signalling.
Circular RNA hsa_circ_0061825 (circ-TFF1) contributes to breast cancer progression through targeting miR-326/TFF1 signalling.
Cell proliferation (2020-01-22)
Gaofeng Pan, Anwei Mao, Jiazhe Liu, Jingfeng Lu, Junbin Ding, Weiyan Liu
ABSTRACT
Circular RNAs (circRNAs) are RNA transcripts that belong to non-coding RNAs (ncRNAs), whose implication in human cancers has been recently demonstrated. However, the specific role of multiple circRNAs in breast cancer remains unidentified. Microarray analysis and bioinformatics analysis were applied to select circRNA and miRNA, respectively. The loop structure of circ-TFF1 was confirmed using RNase R treatment, divergent primer PCR and Sanger sequencing. qRT-PCR and Western blot were employed for gene expressions. In vitro and in vivo experiments were conducted to assess the function of circ-TFF1 in biological processes in breast cancer cells. FISH and subcellular separation indicated circ-TFF1 cellular distribution. Luciferase reporter and RIP assays and Pearson's correlation analysis were performed to evaluate relationships between genes. Circ-TFF1 and TFF1 were both upregulated and positively associated with each other in breast cancer. Knockdown of circ-TFF1 hindered breast cancer cell proliferation, migration, invasion and EMT in vitro and controlled tumour growth in vivo. Circ-TFF1 acted as a ceRNA of TFF1 by sponging miR-326, and its contribution to breast cancer progression was mediated by miR-326/TFF1 axis. Circ-TFF1 is a facilitator in breast cancer relying on TFF1 by absorbing miR-326, providing a novel promising target for BC treatment.
Product Description
Roche
In Situ Cell Death Detection Kit, POD, sufficient for ≤50 tests
Sigma-Aldrich
IgG, Goat, Goat IgG Polyclonal Antibody control validated for use in ELISA.