Merck
  • TRPC1 participates in the HSV-1 infection process by facilitating viral entry.
TRPC1 participates in the HSV-1 infection process by facilitating viral entry.
Science advances (2020-03-25)
DongXu He, AiQin Mao, YouRan Li, SiuCheung Tam, YongTang Zheng, XiaoQiang Yao, Lutz Birnbaumer, Indu S Ambudkar, Xin Ma
ABSTRACT
Mammalian transient receptor potential (TRP) channels are major components of Ca2+ signaling pathways and control a diversity of physiological functions. Here, we report a specific role for TRPC1 in the entry of herpes simplex virus type 1 (HSV-1) into cells. HSV-1-induced Ca2+ release and entry were dependent on Orai1, STIM1, and TRPC1. Inhibition of Ca2+ entry or knockdown of these proteins attenuated viral entry and infection. HSV-1 glycoprotein D interacted with the third ectodomain of TRPC1, and this interaction facilitated viral entry. Knockout of TRPC1 attenuated HSV-1-induced ocular abnormality and morbidity in vivo in TRPC1-/- mice. There was a strong correlation between HSV-1 infection and plasma membrane localization of TRPC1 in epithelial cells within oral lesions in buccal biopsies from HSV-1-infected patients. Together, our findings demonstrate a critical role for TRPC1 in HSV-1 infection and suggest the channel as a potential target for anti-HSV therapy.
Sigma-Aldrich
MISSION® esiRNA, targeting human TRPC1
Pricing and availability is not currently available.