Merck
  • Suppression of beta-N-acetylglucosaminidase in the N-glycosylation pathway for complex glycoprotein formation in Drosophila S2 cells.
Suppression of beta-N-acetylglucosaminidase in the N-glycosylation pathway for complex glycoprotein formation in Drosophila S2 cells.
Glycobiology (2008-12-05)
Yeon Kyu Kim, Kyoung Ro Kim, Dong Gyun Kang, So Young Jang, Young Hwan Kim, Hyung Joon Cha
ABSTRACT
Most insect cells have a simple N-glycosylation process and consequently paucimannosidic or simple core glycans predominate. Previously, we have shown that paucimannosidic N-glycan structures are dominant in Drosophila S2 cells. It has been proposed that beta-N-acetylglucosaminidase (GlcNAcase), a hexosaminidase in the Golgi membrane which removes a terminal N-acetylglucosamine (GlcNAc), might contribute to simple N-glycosylation in several insects and insect-derived cells except S2 cells. In the present work, we investigated the substantial effects of GlcNAcase on N-glycan patterns in Drosophila S2 cells using two GlcNAcase suppression strategies: an mRNA-targeting approach using RNA interference (RNAi) and a protein-targeting approach using the specific chemical inhibitor 2-acetamido-1,2-dideoxynojirimycin (2-ADN). Using high-performance liquid chromatography (HPLC) and matrix-assisted laser desorption/ionization time-of-flight mass spectrometry (MALDI-TOF MS) analyses, we found that the N-glycosylation patterns of human erythropoietin (hEPO) secreted by stably transfected S2 cells were more complex following GlcNAcase suppression, which generated N-glycan structures with a terminal GlcNAc and/or galactose. These data demonstrate that GlcNAcase may be an important factor in the formation of paucimannosidic core N-glycans in Drosophila S2 cells and suggest that it may be possible to express complex glycoproteins in engineered Drosophila S2 cells by suppressing GlcNAcase in the N-glycosylation pathway.