Merck
A versatile route to L-hexoses: synthesis of L-mannose and L-altrose.
Organic letters (2006-10-06)
Annalisa Guaragna, Carmela Napolitano, Daniele D'Alonzo, Silvana Pedatella, Giovanni Palumbo
ABSTRACT
[reaction: see text] An efficient route for the synthesis of orthogonally protected l-sugars has been opened up, starting from the heterocyclic homologating agent 1 and 2,3-O-isopropylidene-l-glyceraldehyde (2). Our synthetic path enables the synthesis of a 2,3-unsaturated-l-pyranoside, which can be suitably functionalized to afford the desired l-hexoses. In this paper, we report the synthesis of l-manno- and l-altro-pyranosides. Moreover, this strategy may be used to prepare all eight sugars and their derivatives in either enantiomeric form.
Sigma-Aldrich
L-(−)-Mannose, ≥99% (GC)