Merck
  • A highly sensitive electrochemical biosensor based on AuNP-modified gold electrodes for selective determination of serum levels of crosslaps.
A highly sensitive electrochemical biosensor based on AuNP-modified gold electrodes for selective determination of serum levels of crosslaps.
3 Biotech (2017-09-29)
Patricia Khashayar, Ghassem Amoabediny, Bagher Larijani, Morteza Hosseini, Rik Verplancke, Michel De Keersmaecker, Annemie Adriaens, Stefan Goemaere, Tom Fiers, Jan Vanfleteren
ABSTRACT
This article explains a step-wise protocol to develop an electrochemical sensor to quantify serum levels of C-telopeptide (CTX) crosslinks also known as crosslaps in a matter of minutes and with high level of accuracy. The technique needs only one-step (incubation) and can thus be used for point of care screening. Due to the excellent electrical properties of the as-prepared immunosensor, CTX levels were successfully measured from 1 to 1000 pg/mL. This is while the normal reference of the marker is 50-450 pg/mL, suggesting that the sensor can acceptably detect CTX. The results also showed a good correlation with ECLIA in measuring serum levels of CTX.
Sigma-Aldrich
Anti-Collagen Type I Antibody, clone C11, ascites fluid, clone C11, Chemicon®