Merck
  • Restriction of HIV-1 Escape by a Highly Broad and Potent Neutralizing Antibody.
Restriction of HIV-1 Escape by a Highly Broad and Potent Neutralizing Antibody.
Cell (2020-02-01)
Philipp Schommers, Henning Gruell, Morgan E Abernathy, My-Kim Tran, Adam S Dingens, Harry B Gristick, Christopher O Barnes, Till Schoofs, Maike Schlotz, Kanika Vanshylla, Christoph Kreer, Daniela Weiland, Udo Holtick, Christof Scheid, Markus M Valter, Marit J van Gils, Rogier W Sanders, Jörg J Vehreschild, Oliver A Cornely, Clara Lehmann, Gerd Fätkenheuer, Michael S Seaman, Jesse D Bloom, Pamela J Bjorkman, Florian Klein
ABSTRACT
Broadly neutralizing antibodies (bNAbs) represent a promising approach to prevent and treat HIV-1 infection. However, viral escape through mutation of the HIV-1 envelope glycoprotein (Env) limits clinical applications. Here we describe 1-18, a new VH1-46-encoded CD4 binding site (CD4bs) bNAb with outstanding breadth (97%) and potency (GeoMean IC50 = 0.048 μg/mL). Notably, 1-18 is not susceptible to typical CD4bs escape mutations and effectively overcomes HIV-1 resistance to other CD4bs bNAbs. Moreover, mutational antigenic profiling uncovered restricted pathways of HIV-1 escape. Of most promise for therapeutic use, even 1-18 alone fully suppressed viremia in HIV-1-infected humanized mice without selecting for resistant viral variants. A 2.5-Å cryo-EM structure of a 1-18-BG505SOSIP.664 Env complex revealed that these characteristics are likely facilitated by a heavy-chain insertion and increased inter-protomer contacts. The ability of 1-18 to effectively restrict HIV-1 escape pathways provides a new option to successfully prevent and treat HIV-1 infection.
Sigma-Aldrich
IgG1, Kappa from human myeloma plasma, purified immunoglobulin, >95% (SDS-PAGE)
Sigma-Aldrich
Nitrilotriacetic acid disodium salt, Sigma Grade, ≥99%
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, 99%
Sigma-Aldrich
IGEPAL® CA-630, for molecular biology
Sigma-Aldrich
N-Hydroxysulfosuccinimide sodium salt, ≥98% (HPLC)