Merck
Tumor-derived CK1α mutations enhance MDMX inhibition of p53.
Oncogene (2019-08-30)
Xia Liu, Qingling Huang, Lihong Chen, Huilai Zhang, Ernst Schonbrunn, Jiandong Chen
ABSTRACT
Somatic missense mutations of the CSNK1A1 gene encoding casein kinase 1 alpha (CK1α) occur in a subset of myelodysplastic syndrome (MDS) with del(5q) karyotype. The chromosomal deletion causes CSNK1A1 haplo-insufficiency. CK1α mutations have also been observed in a variety of solid and hematopoietic tumors at low frequency. The functional consequence of CK1α mutation remains unknown. Here we show that tumor-associated CK1α mutations exclusively localize to the substrate-binding cleft. Functional analysis of recurrent mutants E98K and D140A revealed enhanced binding to the p53 inhibitor MDMX, increased ability to stimulate MDMX-p53 binding, and increased suppression of p21 expression. Furthermore, E98K and D140A mutants have reduced ability to promote phosphorylation of β-catenin, resulting in enhanced Wnt signaling. The results suggest that the CK1α mutations observed in tumors cause gain-of-function in cooperating with MDMX and inhibiting p53, and partial loss-of-function in suppressing Wnt signaling. These functional changes may promote expansion of abnormal myeloid progenitors in del(5q) MDS, and in rare cases drive the progression of other tumors.
Sigma-Aldrich
MISSION® esiRNA, targeting human CSNK1A1, CTB-89H12.4
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid