Merck
Dorsal tegmental dopamine neurons gate associative learning of fear.
Nature neuroscience (2018-06-29)
Florian Groessl, Thomas Munsch, Susanne Meis, Johannes Griessner, Joanna Kaczanowska, Pinelopi Pliota, Dominic Kargl, Sylvia Badurek, Klaus Kraitsy, Arash Rassoulpour, Johannes Zuber, Volkmar Lessmann, Wulf Haubensak
ABSTRACT
Functional neuroanatomy of Pavlovian fear has identified neuronal circuits and synapses associating conditioned stimuli with aversive events. Hebbian plasticity within these networks requires additional reinforcement to store particularly salient experiences into long-term memory. Here we have identified a circuit that reciprocally connects the ventral periaqueductal gray and dorsal raphe region with the central amygdala and that gates fear learning. We found that ventral periaqueductal gray and dorsal raphe dopaminergic (vPdRD) neurons encode a positive prediction error in response to unpredicted shocks and may reshape intra-amygdala connectivity via a dopamine-dependent form of long-term potentiation. Negative feedback from the central amygdala to vPdRD neurons might limit reinforcement to events that have not been predicted. These findings add a new module to the midbrain dopaminergic circuit architecture underlying associative reinforcement learning and identify vPdRD neurons as a critical component of Pavlovian fear conditioning. We propose that dysregulation of vPdRD neuronal activity may contribute to fear-related psychiatric disorders.
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, Chemicon®, from chicken