Merck
  • ARHGAP10 Inhibits the Proliferation and Metastasis of CRC Cells via Blocking the Activity of RhoA/AKT Signaling Pathway.
ARHGAP10 Inhibits the Proliferation and Metastasis of CRC Cells via Blocking the Activity of RhoA/AKT Signaling Pathway.
OncoTargets and therapy (2020-01-11)
Lin Liu, Dongyu Xie, Haina Xie, Wei Huang, Jingxian Zhang, Wenye Jin, Wenjing Jiang, Donghao Xie
ABSTRACT
ARHGAP10 belongs to the ARHGAP family, which is downregulated in certain human tumors. However, the detailed function of ARHGAP10 remains unclear in human colon carcinoma (CRC). In the current study, we aimed to explore the role of ARHGAP10 in the growth and metastasis of CRC cells. ARHGAP10 was induced silencing and overexpression using RNA interference (RNAi) and lentiviral-vector in CRC cells. Quantitative real-time PCR (qRT-PCR) and Western blot were used to quantify the mRNA and protein contents of ARHGAP10. Cell proliferation was determined by using Cell counting kit-8 (CCK-8). Transwell assay was utilized to examine the role of ARHGAP10 in the migration and invasion of CRC cells. Our results indicated that ARHGAP10 was downregulated in human CRC tissues and low expression of ARHGAP10 was associated with poor prognosis of patients with CRC. Moreover, ARHGAP10 overexpression significantly inhibited the proliferation and metastasis of CRC cells. Moreover, a PI3K/AKT inhibitor LY294002 was utilized to examine the connection between ARHGAP10 and AKT. Our findings demonstrated that the AKT inhibitor LY294002 could rescue the function of ARHGAP10 in CRC cells. It was the first time to elucidate that AKT involved in the ARHGAP10 signaling pathway and ARHGAP10 negatively mediated the phosphorylation of AKT (p-AKT) and RhoA activity in CRC cells. Interestingly, the Rho/MRTF/SRF inhibitor CCG-1423 significantly inhibited the phosphorylation of AKT in ARHGAP10 siRNA transfected CRC cells. Much importantly, overexpression of ARHGAP10 deeply suppressed the metastasis of CRC cells in the lung in vivo. Taken together, our findings not only enhanced the understanding of the anti-cancer effect of ARHGAP10 in CRC cells but also indicated its underlying pathway in CRC.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Pricing and availability is not currently available.
Millipore
Immobilon®-NC Transfer Membrane, 1 roll, 33 cm x 3 m, 0.45 µm pore size, Triton-free, mixed cellulose esters transfer membrane
Pricing and availability is not currently available.