Merck
  • Cyclooxygenase-2 (COX-2) inhibition constrains indoleamine 2,3-dioxygenase 1 (IDO1) activity in acute myeloid leukaemia cells.
Cyclooxygenase-2 (COX-2) inhibition constrains indoleamine 2,3-dioxygenase 1 (IDO1) activity in acute myeloid leukaemia cells.
Molecules (Basel, Switzerland) (2013-08-27)
Maria Grazia Iachininoto, Eugenia Rosa Nuzzolo, Giuseppina Bonanno, Andrea Mariotti, Annabella Procoli, Franco Locatelli, Raimondo De Cristofaro, Sergio Rutella
ABSTRACT
Indoleamine 2,3-dioxygenase 1 (IDO1) metabolizes L-tryptophan to kynurenines (KYN), inducing T-cell suppression either directly or by altering antigen-presenting-cell function. Cyclooxygenase (COX)-2, the rate-limiting enzyme in the synthesis of prostaglandins, is over-expressed by several tumours. We aimed at determining whether COX-2 inhibitors down-regulate the IFN-g-induced expression of IDO1 in acute myeloid leukaemia (AML) cells. IFN-γ at 100 ng/mL up-regulated COX-2 and IDO1 in HL-60 AML cells, both at mRNA and protein level. The increased COX-2 and IDO1 expression correlated with heightened production of prostaglandin (PG)E₂ and kynurenines, respectively. Nimesulide, a preferential COX-2 inhibitor, down-regulated IDO1 mRNA/protein and attenuated kynurenine synthesis, suggesting that overall IDO inhibition resulted both from reduced IDO1 gene transcription and from inhibited IDO1 catalytic activity. From a functional standpoint, IFN-g-challenged HL-60 cells promoted the in vitro conversion of allogeneic CD4⁺CD25⁻ T cells into bona fide CD4⁺CD25⁺FoxP3⁺ regulatory T cells, an effect that was significantly reduced by treatment of IFN-γ-activated HL-60 cells with nimesulide. Overall, these data point to COX-2 inhibition as a potential strategy to be pursued with the aim at circumventing leukaemia-induced, IDO-mediated immune dysfunction.
Sigma-Aldrich
Anti-Indoleamine 2,3-dioxygenase Antibody, clone 10.1, clone 10.1, Chemicon®, from mouse