Merck
  • Secretory IgA from submucosal glands does not compensate for its airway surface deficiency in chronic obstructive pulmonary disease.
Secretory IgA from submucosal glands does not compensate for its airway surface deficiency in chronic obstructive pulmonary disease.
Virchows Archiv : an international journal of pathology (2015-10-04)
Rui-Hong Du, Bradley W Richmond, Timothy S Blackwell, Justin M Cates, Pierre P Massion, Lorraine B Ware, Jae Woo Lee, Alexey V Kononov, William E Lawson, Timothy S Blackwell, Vasiliy V Polosukhin
ABSTRACT
Secretory immunoglobulin A (SIgA) reaches the airway lumen by local transcytosis across airway epithelial cells or with tracheobronchial submucosal gland secretions. In chronic obstructive pulmonary disease (COPD), deficiency of SIgA on the airway surface has been reported. However, reduction of SIgA levels in sputum and bronchoalveolar lavage (BAL) fluid has not been consistently observed. To explain this discrepancy, we analyzed BAL fluid and lung tissue from patients with COPD and control subjects. Immunohistochemical analysis of large and small airways of COPD patients showed that MUC5AC is the predominant mucin expressed by airway epithelial cells, whereas MUC5B is expressed in submucosal glands of large airways. Dual immunostaining with anti-IgA and anti-MUC5B antibodies showed reduction of IgA on the airway surface as well as accumulation of IgA within MUC5B-positive luminal mucus plugs, suggesting that luminal SIgA originates from submucosal glands in COPD patients. We found that the concentration of SIgA in BAL is inversely correlated with forced expiratory volume in 1 s (FEV1) in COPD, but that the ratio of SIgA/MUC5B is a better predictor of FEV1, particularly in patients with moderate COPD. Together, these findings suggest that SIgA production by submucosal glands, which are expanded in COPD, is insufficient to compensate for reduced SIgA transcytosis by airway epithelial cells. Localized SIgA deficiency on the surface of small airways is associated with COPD progression and represents a potential new therapeutic target in COPD.
Sigma-Aldrich
Anti-Mucin MUC5AC Antibody, clone CLH2, clone CLH2, Chemicon®, from mouse