Merck
  • GIGYF1/2-Driven Cooperation between ZNF598 and TTP in Posttranscriptional Regulation of Inflammatory Signaling.
GIGYF1/2-Driven Cooperation between ZNF598 and TTP in Posttranscriptional Regulation of Inflammatory Signaling.
Cell reports (2019-03-28)
Maxim A X Tollenaere, Christopher Tiedje, Simon Rasmussen, Julie C Nielsen, Anna C Vind, Melanie Blasius, Tanveer S Batth, Niels Mailand, Jesper V Olsen, Matthias Gaestel, Simon Bekker-Jensen
ABSTRACT
Inflammatory signaling is restricted through degradation and the translational repression of cytokine mRNAs. A key factor in this regulation is tristetraprolin (TTP), an RNA-binding protein (RBP) that recruits RNA-destabilizing factors and the translation inhibitory complex 4EHP-GIGYF1/2 to AU-rich element (ARE)-containing mRNAs. Here, we show that the RBP ZNF598 contributes to the same regulatory module in a TTP-like manner. Similar to TTP, ZNF598 harbors three proline-rich motifs that bind the GYF domain of GIGYF1. RNA sequencing experiments showed that ZNF598 is required for the regulation of known TTP targets, including IL-8 and CSF2 mRNA. Furthermore, we demonstrate that ZNF598 binds to IL-8 mRNA, but not TNF mRNA. Collectively, our findings highlight that ZNF598 functions as an RBP that buffers the level of a range of mRNAs. We propose that ZNF598 is a TTP-like factor that can contribute to the regulation of the inflammatory potential of cytokine-producing cells.
Sigma-Aldrich
Anti-TTP (N-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)
Sigma-Aldrich
Anti-ZNF598 antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)