Merck
  • Illicit Upregulation of Serotonin Signaling Pathway in Adrenals of Patients With High Plasma or Intra-Adrenal ACTH Levels.
Illicit Upregulation of Serotonin Signaling Pathway in Adrenals of Patients With High Plasma or Intra-Adrenal ACTH Levels.
The Journal of clinical endocrinology and metabolism (2019-05-11)
Julie Le Mestre, Céline Duparc, Yves Reznik, Fidéline Bonnet-Serrano, Philippe Touraine, Olivier Chabre, Jacques Young, Mari Suzuki, Mathilde Sibony, Françoise Gobet, Constantine A Stratakis, Gérald Raverot, Jérôme Bertherat, Hervé Lefebvre, Estelle Louiset
ABSTRACT
In the human adrenal, serotonin (5-HT), released by mast cells stimulates corticosteroid secretion through activation of type 4 serotonin receptors (5-HT4R). In primary pigmented nodular adrenocortical disease cells, activation of the cAMP/protein kinase A (PKA) pathway by PRKAR1A mutations triggers upregulation of the 5-HT synthesizing enzyme tryptophan hydroxylase (TPH) and the 5-HT4, 5-HT6, and 5-HT7 receptors. Because ACTH stimulates cortisol secretion through activation of PKA, adrenocortical tissues exposed to sustained stimulation by ACTH may harbor increased expression of TPH and 5-HT4/6/7 receptors. To investigate the effects of long-term ACTH stimulation on the serotonergic pathway in adrenals of patients with high plasma or intra-adrenal ACTH levels. Adrenal tissues were obtained from patients with Cushing disease, ectopic secretion of ACTH [paraneoplastic Cushing syndrome; (paraCS)], 21-hydroxylase deficiency (21-OHD), primary bilateral macronodular adrenal hyperplasia with intra-adrenal ACTH presence, or cortisol-producing adenomas. TPH and 5-HT4/6/7 receptor expression was investigated using RT-PCR and immunochemistry in comparison with normal adrenals. Primary cultured adrenocortical cells originating from a patient with paraCS were incubated with 5-HT and 5-HTR agonists/antagonists. TPH and/or 5-HT4/6/7 receptors were overexpressed in the different types of tissues. In paraCS cultured cells, the cortisol response to 5-HT was exaggerated compared with normal adrenal cells and the stimulatory action of 5-HT was reduced by 5-HT4R antagonist. Our results indicate that prolonged activation of the cAMP/PKA pathway by ACTH induces an aberrant serotonergic stimulatory loop in the adrenal cortex that likely participates in the pathogenesis of corticosteroid hypersecretion.
Sigma-Aldrich
Anti-TPH1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution