Merck
Lysine methylation promotes VEGFR-2 activation and angiogenesis.
Science signaling (2013-12-05)
Edward J Hartsough, Rosana D Meyer, Vipul Chitalia, Yan Jiang, Victor E Marquez, Irina V Zhdanova, Janice Weinberg, Catherine E Costello, Nader Rahimi
ABSTRACT
Activation of vascular endothelial growth factor receptor-2 (VEGFR-2), an endothelial cell receptor tyrosine kinase, promotes tumor angiogenesis and ocular neovascularization. We report the methylation of VEGFR-2 at multiple Lys and Arg residues, including Lys(1041), a residue that is proximal to the activation loop of the kinase domain. Methylation of VEGFR-2 was independent of ligand binding and was not regulated by ligand stimulation. Methylation of Lys(1041) enhanced tyrosine phosphorylation and kinase activity in response to ligands. Additionally, interfering with the methylation of VEGFR-2 by pharmacological inhibition or by site-directed mutagenesis revealed that methylation of Lys(1041) was required for VEGFR-2-mediated angiogenesis in zebrafish and tumor growth in mice. We propose that methylation of Lys(1041) promotes the activation of VEGFR-2 and that similar posttranslational modification could also regulate the activity of other receptor tyrosine kinases.
Sigma-Aldrich
Anti-phospho-KDR/Flk-1/VEGFR2 (Tyr1054) Antibody, clone D1W, rabbit monoclonal, culture supernatant, clone D1W, Upstate®
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse