Merck
  • Biocompatible Inhibitor Based on Chitosan and Amphiphilic Peptide against Mutant Huntingtin Toxicity.
Biocompatible Inhibitor Based on Chitosan and Amphiphilic Peptide against Mutant Huntingtin Toxicity.
Chembiochem : a European journal of chemical biology (2019-06-06)
Devi Wahyuningtyas, Wen-Hao Chen, Chen-Han Huang, Yu-Jung He, Joseph Jen-Tse Huang
ABSTRACT
Huntington's disease (HD) is classified as a protein-misfolding disease correlated with the mutant Huntingtin (mHtt) protein with abnormally expanded polyglutamine (polyQ) domains. Because no effective drugs have yet been reported, attempts to develop better therapy to delay the age of onset are in urgent demand. In this study, an amphiphilic peptide consisting of negatively charged hexaglutamic acid and a stretch of decaglutamine (E6 Q10 ) was chemically synthesized as an inhibitor against polyQ and mHtt toxicity. It is found that E6 Q10 selfassembles into spherical vesicles, as shown by means of TEM, cryoelectron microscopy, and dynamic light scattering. Assembled E6 Q10 prevented the polyQ-rich peptide (KKWQ20 AKK) from forming amyloid fibrils. To enable the cell-penetration ability of E6 Q10 , the E6 Q10 ⋅chitosan complex was generated. It is demonstrated that the complex penetrates cells, interferes with the mHtt oligomerization and aggregation process, and prevents mHtt cytotoxicity. By combining positively charged chitosan and amphiphilic peptides with a negatively charge moiety, a new strategy is provided to develop biocompatible and biodegradable inhibitors against mHtt toxicity.
Sigma-Aldrich
1,1,1,3,3,3-Hexafluoro-2-propanol, ≥99%
Sigma-Aldrich
Fmoc-Glu(OtBu)-OH, ≥98.0% (HPLC)
Sigma-Aldrich
Fmoc-Ala-OH, 95%
Sigma-Aldrich
Triisopropylsilane, 98%