Merck
  • Short-term clinical and immunologic effects of poly-gamma-glutamic acid (γ-PGA) in women with cervical intraepithelial neoplasia 1 (CIN 1): A multicenter, randomized, double blind, phase II trial.
Short-term clinical and immunologic effects of poly-gamma-glutamic acid (γ-PGA) in women with cervical intraepithelial neoplasia 1 (CIN 1): A multicenter, randomized, double blind, phase II trial.
PloS one (2019-06-21)
Hyun-Woong Cho, Young-Chul Park, Moon-Hee Sung, Jong Sup Park, Tae Jin Kim, Seok Ju Seong, Chi Heum Cho, Jae Kwan Lee
ABSTRACT
The aim of this study was to investigate the short-term efficacy and safety of Poly-gamma-glutamic acid (γ-PGA) and the immunologic changes in patients with CIN 1. Participants were randomly assigned to one of two groups and orally treated with placebo or 1,500 mg of γ-PGA for 4 weeks. The primary endpoint of the study was histologic regression rate of CIN 1 at 12 weeks between γ-PGA and control groups. The secondary endpoints were HPV clearance and change in immune responses. From April 2013 to December 2015, 195 patients participated in the study. In the intention-to-treat analysis, 42 (42.4%) of the women who received γ-PGA experienced histologic remission versus 26 (27.1%) in the control group, with a statistically significant difference (p = 0.018). In the γ-PGA group, HPV clearance was found in 37 (43.5%) of 85 patients infected with high-risk HPV, showing a significant difference compared to the control group, in which 20 (26.7%) of 75 patients exhibited HPV clearance (p = 0.026). However, there was no significant difference between the two groups in the change of NK cell activity, major histocompatibility complex (MHC) class II CD8 count, and CD56 count. γ-PGA showed a short-term therapeutic effect on CIN 1 and high-risk HPV infection. It is a non-invasive, promising oral medication for women with these conditions. Clinical Trials NCT01826045.
Sigma-Aldrich
Boc-Leu-OH hydrate, ≥99.0% (HPLC)
Sigma-Aldrich
N,N′-Dimethylurea, (sym.), ≥99% (from N)