Merck
  • Synthesis, Biological Evaluation and Structure-Activity Relationships of Diflapolin Analogues as Dual sEH/FLAP Inhibitors.
Synthesis, Biological Evaluation and Structure-Activity Relationships of Diflapolin Analogues as Dual sEH/FLAP Inhibitors.
ACS medicinal chemistry letters (2019-01-19)
Lisa Vieider, Erik Romp, Veronika Temml, Jana Fischer, Christian Kretzer, Martin Schoenthaler, Abdulla Taha, Victor Hernández-Olmos, Sonja Sturm, Daniela Schuster, Oliver Werz, Ulrike Garscha, Barbara Matuszczak
ABSTRACT
A series of derivatives of the potent dual soluble epoxide hydrolase (sEH)/5-lipoxygenase-activating protein (FLAP) inhibitor diflapolin was designed, synthesized, and characterized by 1H NMR, 13C NMR, and elemental analysis. These novel compounds were biologically evaluated for their inhibitory activity against sEH and FLAP. Molecular modeling tools were applied to analyze structure-activity relationships (SAR) on both targets. Results show that even small modifications on the lead compound diflapolin markedly influence the inhibitory potential, especially on FLAP, suggesting very narrow SAR.
Sigma-Aldrich
Diflapolin, ≥98% (HPLC)