Merck
Cellular mechanisms of beta-amyloid production and secretion.
Proceedings of the National Academy of Sciences of the United States of America (1999-09-29)
S Sinha, I Lieberburg
ABSTRACT
The major constituent of senile plaques in Alzheimer's disease is a 42-aa peptide, referred to as beta-amyloid (Abeta). Abeta is generated from a family of differentially spliced, type-1 transmembrane domain (TM)-containing proteins, called APP, by endoproteolytic processing. The major, relatively ubiquitous pathway of APP metabolism in cell culture involves cleavage by alpha-secretase, which cleaves within the Abeta sequence, thus precluding Abeta formation and deposition. An alternate secretory pathway, enriched in neurons and brain, leads to cleavage of APP at the N terminus of the Abeta peptide by beta-secretase, thus generating a cell-associated beta-C-terminal fragment (beta-CTF). A pathogenic mutation at codons 670/671 in APP (APP "Swedish") leads to enhanced cleavage at the beta-secretase scissile bond and increased Abeta formation. An inhibitor of vacuolar ATPases, bafilomycin, selectively inhibits the action of beta-secretase in cell culture, suggesting a requirement for an acidic intracellular compartment for effective beta-secretase cleavage of APP. beta-CTF is cleaved in the TM domain by gamma-secretase(s), generating both Abeta 1-40 (90%) and Abeta 1-42 (10%). Pathogenic mutations in APP at codon 717 (APP "London") lead to an increased proportion of Abeta 1-42 being produced and secreted. Missense mutations in PS-1, localized to chromosome 14, are pathogenic in the majority of familial Alzheimer's pedigrees. These mutations also lead to increased production of Abeta 1-42 over Abeta 1-40. Knockout of PS-1 in transgenic animals leads to significant inhibition of production of both Abeta 1-40 and Abeta 1-42 in primary cultures, indicating that PS-1 expression is important for gamma-secretase cleavages. Peptide aldehyde inhibitors that block Abeta production by inhibiting gamma-secretase cleavage of beta-CTF have been discovered.
Sigma-Aldrich
Bafilomycin A1, InSolution, ≥90%, A macrolide antibiotic that acts as a specific inhibitor of vacuolar-type H+-ATPase
Pricing and availability is not currently available.