Merck
  • Development of immortalized Hertwig's epithelial root sheath cell lines for cementum and dentin regeneration.
Development of immortalized Hertwig's epithelial root sheath cell lines for cementum and dentin regeneration.
Stem cell research & therapy (2019-01-05)
Xuebing Li, Sicheng Zhang, Zirui Zhang, Weihua Guo, Guoqing Chen, Weidong Tian
ABSTRACT
Hertwig's epithelial root sheath (HERS) is important in guiding tooth root formation by differentiating into cementoblasts through epithelial-mesenchymal transition (EMT) and inducing odontoblastic differentiation of dental papilla through epithelial-mesenchymal interaction (EMI) during the tooth root development. Thus, HERS cells are critical for cementum and dentin formation and might be a potential cell source to achieve tooth root regeneration. However, limited availability and lifespan of primary HERS cells may represent an obstacle for biological investigation and therapeutic use of tooth tissue engineering. Therefore, we constructed, characterized, and tested the functionality of immortalized cell lines in order to produce a more readily available alternative to HERS cells. Primary HERS cells were immortalized via infection with lentivirus vector containing the gene encoding simian virus 40 Large T Antigen (SV40LT). Immortalized HERS cell subclones were isolated using a limiting dilution method, and subclones named HERS-H1 and HERS-C2 cells were isolated. The characteristics of HERS-H1 and HERS-C2 cells, including cell proliferation, ability of epithelial-mesenchymal transformation and epithelial-mesenchymal interaction, were determined by CCK-8 assay, immunofluorescence staining, and real-time PCR. The cell differentiation into cementoblast-like cells or periodontal fibroblast-like cells was confirmed in vivo. And the inductive influence of the cell lines on dental papilla cells (DPCs) was also confirmed in vivo. HERS-H1 and HERS-C2 cells share some common features with primary HERS cells such as epithelial-like morphology, positive expression of CK14, E-Cadherin, and Vimentin, and undergoing EMT in response to TGF-beta. HERS-C2 cells showed the EMT characteristics and could differentiate into cementum-forming cells in vitro and generate cementum-like tissue in vivo. HERS-H1 could induce the differentiation of DPCs into odontoblasts in vitro and generation of dentin-like tissue in vivo. We successfully isolated and characterized novel cell lines representing two key features of HERS cells during the tooth root development and which were useful substitutes for primary HERS cells, thereby providing a biologically relevant, unlimited cell source for studies on cell biology, developmental biology, and tooth root regeneration.
Sigma-Aldrich
Anti-Cytokeratin 14 Antibody, clone RCK107, clone RCK107, Chemicon®, from mouse