- Effects of processing and storage conditions on volatile composition and odor characteristics of blackcurrant (Ribes nigrum) juices.
Effects of processing and storage conditions on volatile composition and odor characteristics of blackcurrant (Ribes nigrum) juices.
Blackcurrant juices were prepared without enzyme (NEB-juice) and with conventional enzyme-aided berry pressing (EB-juice). Juices were pasteurized and stored at ambient temperature (in light and dark conditions) and at +4 °C for a 1-year period of time. Volatile composition and odor attributes were followed by HS-SPME-GC-MS and a sensory panel, respectively. Volatiles were decreased in the EB-juice, showing 100-fold lower contents of the main terpenoids, e.g., α-pinene, δ-3-carene, limonene, terpinolene, bornyl acetate and β-caryophyllene; whereas esters, such as methyl butanoate and ethyl butanoate, showed 2- and 4-fold lower contents for the EB-juice. Pasteurization produced minor changes in both juices. Volatiles decreased during storage at room temperature while at +4 °C esters and eucalyptol were kept at 100%. The descriptive sensory analysis reported a significant decrease (p < 0.05) of the berry-like odor after 12-month storage at RT. Storage at +4 °C for 12 months did not affect the odor quality.