Merck
  • Effect of Dapagliflozin Treatment on the Expression of Renal Sodium Transporters/Channels on High-Fat Diet Diabetic Mice.
Effect of Dapagliflozin Treatment on the Expression of Renal Sodium Transporters/Channels on High-Fat Diet Diabetic Mice.
Nephron (2019-02-26)
Chao Ma, Jeroen H F de Baaij, Paul J Millar, Victor A Gault, Bastiaan E de Galan, René J M Bindels, Joost G J Hoenderop
ABSTRACT
Inhibition of the Na+/glucose co-transporter 2 is a new therapeutic strategy for diabetes. It is unclear how proximal loss of Na+ (and glucose) affects the subsequent Na+ transporters in the proximal tubule (PT), thick ascending limb of loop of Henle (TAL), distal convoluted tubule (DCT) and collecting duct (CD). Mice on a high fat diet were administered 3 doses streptozotocin 6 days prior to oral dapagliflozin administration or vehicle for 18 days. A control group of lean mice were also included. Body weight and glucose were recorded at regular intervals during treatment. Renal Na+ transporters expression in nephron segments were analyzed by RT-qPCR and Western blot. Dapagliflozin treatment resulted in a significant reduction in body weight and blood glucose compared to vehicle-treated controls. mRNA results showed that Na+-hydrogen antiporter 3 (NHE3), Na+/phosphate cotransporter (NaPi-2a) and epithelial Na+ channel expression was increased, Ncx1, ENaCβ and ENaCγ expression declined (p all < 0.05), respectively, in dapagliflozin-treated mice when compared with saline vehicle mice. Na-K-2Cl cotransporters and Na-Cl cotransporter mRNA expression was not affected by dapagliflozin treatment. Na+/K+-ATPase (Atp1b1) expression was also increased significantly by dapagliflozin treatment, but it did not affect Atp1a1 and glucose transporter 2 expression. Western blot analysis showed that NaPi-2a, NHE3 and ATP1b1 expression was upregulated in dapagliflozin-treated diabetic mice when compared with saline vehicle mice (p < 0.05). Our findings suggest that dapagliflozin treatment augments compensatory changes in the renal PT in diabetic mice.