Merck
  • Involvement of the Adhesion GPCRs Latrophilins in the Regulation of Insulin Release.
Involvement of the Adhesion GPCRs Latrophilins in the Regulation of Insulin Release.
Cell reports (2019-02-07)
Juliane Röthe, Doreen Thor, Jana Winkler, Alexander B Knierim, Claudia Binder, Sandra Huth, Robert Kraft, Sven Rothemund, Torsten Schöneberg, Simone Prömel
ABSTRACT
Insulin secretion from pancreatic β cells is a highly complex and tightly regulated process. Its dysregulation is one characteristic of type 2 diabetes, and thus, an in-depth understanding of the mechanisms controlling insulin secretion is essential for rational therapeutic intervention. G-protein-coupled receptors (GPCRs) have been established as major regulators of insulin exocytosis. Recent studies also suggest the involvement of adhesion GPCRs, a non-prototypical class of GPCRs. Here, we identify latrophilins, which belong to the class of adhesion GPCRs, to be highly expressed in different cell types of pancreatic islets. In vitro and ex vivo analyses show that distinct splice variants of the latrophilin LPHN3/ADGRL3 decrease insulin secretion from pancreatic β cells by reducing intracellular cyclic AMP levels via the Gi-mediated pathway. Our data highlight the key role of LPHN3 in modulating insulin secretion and its potential as therapeutic target for type 2 diabetes.
Product Description
Roche
Anti-HA-Peroxidase, High Affinity, from rat IgG1
Sigma-Aldrich
Anti-Actin (Ab-1) Mouse mAb (JLA20), liquid, clone JLA20, Calbiochem®