Merck
  • A Free Radical-Generating System Regulates Amyloid Oligomers: Involvement of Cathepsin B.
A Free Radical-Generating System Regulates Amyloid Oligomers: Involvement of Cathepsin B.
Journal of Alzheimer's disease : JAD (2018-11-08)
Patricia Llorente, Henrike Kristen, Isabel Sastre, Ana Toledano-Zaragoza, Jesús Aldudo, María Recuero, María J Bullido
ABSTRACT
Amyloid-β (Aβ), a major component of senile plaques, is generated via the proteolysis of amyloid-β protein precursor (AβPP). This cleavage also produces AβPP fragment-derived oligomers which can be highly neurotoxic. AβPP metabolism/processing is affected by many factors, one of which is oxidative stress (OS). Associated with aging, OS is an important risk factor for Alzheimer's disease. In addition, the protein degradation systems, especially those involving cathepsins, are impaired in aging brains. Moreover, cathepsin B (CTSB) is a cysteine protease with potentially specific roles in AβPP proteolysis (β-secretase activity) and Aβ clearance (Aβ degradative activity). The present work examines the effect of OS and the involvement of CTSB in amyloid oligomer formation. The xanthine/xanthine oxidase (X-XOD) free radical generating system induced the partial inhibition of CTSB activity, which was accompanied by an increase in large amyloid oligomers. These were located throughout the cytosol and in endo-lysosomal vesicles. Cells treated with the CTSB inhibitor CA-074Me also showed increased amyloid oligomer levels, whereas those subjected to OS in the presence of the inhibitor showed no such increase. However, CTSB inhibition clearly modulated the AβPP metabolism/processing induced by X-XOD, as revealed by the increase in intracellular AβPP and secreted α-secretase-cleaved soluble AβPP. The present results suggest that CTSB participates in the changes of amyloid oligomer induced by mild OS.