Merck
  • Knockdown of KDM1B inhibits cell proliferation and induces apoptosis of pancreatic cancer cells.
Knockdown of KDM1B inhibits cell proliferation and induces apoptosis of pancreatic cancer cells.
Pathology, research and practice (2019-03-09)
Yun Wang, Liankang Sun, Yumei Luo, Shuixiang He
ABSTRACT
Pancreatic cancer (PC) is one of the common malignant tumors in digestive tract with a high fatality rate. The oncogenic role of lysine-specific demethylase1 (LSD1/KDM1 A) has been well recognized in PC. While, the role of its homolog LSD2 (KDM1B) in regulating PC progression is poorly understood. In this study, we attempted to evaluate the functional role of KDM1B in PC cells. The expression of KDM1B was detected by immunohistochemistry and immunoblotting in PC tissues and cells. Lentivirus-mediated shRNA was applied to silence KDM1B in PANC-1 and SW1990 cells. Cell proliferation was measured by MTT and Celigo assay. Cell apoptosis was determined by both Caspase-Glo®3/7 assay and Flow cytometry. Intracellular signaling molecules were detected using a PathScan intracellular signaling array kit. In this study, we found KDM1B was highly expressed in PC tissues compared to paracancerous tissues. Moreover, elevated expression of KDM1B was detected in PC cell lines (BxPC-3, CFPAC-1, PANC-1 and SW1990) as compared with a normal human pancreatic duct epithelial cell line (HPDE6-C7). Further investigations revealed that KDM1B knockdown significantly inhibited PC cell proliferation. Furthermore, the apoptosis of PANC-1 and SW1990 cells was significantly increased after KDM1B knockdown. Notably, the activations of p-ERK1/2, p-Smad2, p-p53, cleaved PARP, cleaved Caspase-3, cleaved Caspase-7, p-eIF2a and Survivin were promoted by KDM1B knockdown, while IkBa was suppressed. Taken together, our findings provided new insights into the critical and multifaceted roles of KDM1B in the regulation of cell proliferation and apoptosis, and offered a potentially novel target in preventing the progression of PC.