Merck
  • Drosophila p53 integrates the antagonism between autophagy and apoptosis in response to stress.
Drosophila p53 integrates the antagonism between autophagy and apoptosis in response to stress.
Autophagy (2018-12-20)
Marion Robin, Abdul Raouf Issa, Cristiana C Santos, Francesco Napoletano, Céline Petitgas, Gilles Chatelain, Mathilde Ruby, Ludivine Walter, Serge Birman, Pedro M Domingos, Brian R Calvi, Bertrand Mollereau
ABSTRACT
The tumor suppressor TP53/p53 is a known regulator of apoptosis and macroautophagy/autophagy. However, the molecular mechanism by which TP53 regulates 2 apparently incompatible processes remains unknown. We found that Drosophila lacking p53 displayed impaired autophagic flux, higher caspase activation and mortality in response to oxidative stress compared with wild-type flies. Moreover, autophagy and apoptosis were differentially regulated by the p53 (p53B) and ΔNp53 (p53A) isoforms: while the former induced autophagy in differentiated neurons, which protected against cell death, the latter inhibited autophagy by activating the caspases Dronc, Drice, and Dcp-1. Our results demonstrate that the differential use of p53 isoforms combined with the antagonism between apoptosis and autophagy ensures the generation of an appropriate p53 biological response to stress.
Product Description
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in a glass vial
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture