Merck
  • Fullerenyl boronic esters: ferric perchlorate-mediated synthesis and functionalization.
Fullerenyl boronic esters: ferric perchlorate-mediated synthesis and functionalization.
Organic letters (2012-03-16)
Fa-Bao Li, Xun You, Tong-Xin Liu, Guan-Wu Wang
ABSTRACT
Fullerenyl boronic esters have been prepared by a ferric perchlorate-promoted reaction of [60]fullerene with various arylboronic acids. The obtained fullerenyl boronic esters could undergo further functionalization with diols to afford C(60)-fused dioxane/dioxepane derivatives. A possible reaction mechanism for the formation of fullerenyl boronic esters has been proposed.
Sigma-Aldrich
4-Cyanophenylboronic acid, ≥95%
Sigma-Aldrich
2-Thienylboronic acid, ≥95.0%