Merck
  • Role of excited states in low-energy electron (LEE) induced strand breaks in DNA model systems: influence of aqueous environment.
Role of excited states in low-energy electron (LEE) induced strand breaks in DNA model systems: influence of aqueous environment.
Chemphyschem : a European journal of chemical physics and physical chemistry (2009-03-25)
Anil Kumar, Michael D Sevilla
ABSTRACT
Low-energy electrons (LEEs) interact with a DNA model system to create a variety of excited states. In the gas phase, dissociative (sigma*) states are accessible by LEEs with energy <4 eV (see picture) and cause facile strand breaks through a dissociative electron attachment mechanism. However, under solvation these dissociative (sigma*) states are blue-shifted and are less accessible to LEEs.
Sigma-Aldrich
Poly(deoxyguanylic-deoxycytidylic) acid sodium salt, double stranded, alternating copolymer