Merck
  • A dynamic view of the proteomic landscape during differentiation of ReNcell VM cells, an immortalized human neural progenitor line.
A dynamic view of the proteomic landscape during differentiation of ReNcell VM cells, an immortalized human neural progenitor line.
Scientific data (2019-02-20)
Yuyu Song, Kartik Subramanian, Matthew J Berberich, Steven Rodriguez, Isabel J Latorre, Catherine M Luria, Robert Everley, Mark W Albers, Timothy J Mitchison, Peter K Sorger
ABSTRACT
The immortalized human ReNcell VM cell line represents a reproducible and easy-to-propagate cell culture system for studying the differentiation of neural progenitors. To better characterize the starting line and its subsequent differentiation, we assessed protein and phospho-protein levels and cell morphology over a 15-day period during which ReNcell progenitors differentiated into neurons, astrocytes and oligodendrocytes. Five of the resulting datasets measured protein levels or states of phosphorylation based on tandem-mass-tag (TMT) mass spectrometry and four datasets characterized cellular phenotypes using high-content microscopy. Proteomic analysis revealed reproducible changes in pathways responsible for cytoskeletal rearrangement, cell phase transitions, neuronal migration, glial differentiation, neurotrophic signalling and extracellular matrix regulation. Proteomic and imaging data revealed accelerated differentiation in cells treated with the poly-selective CDK and GSK3 inhibitor kenpaullone or the HMG-CoA reductase inhibitor mevastatin, both of which have previously been reported to promote neural differentiation. These data provide in-depth information on the ReNcell progenitor state and on neural differentiation in the presence and absence of drugs, setting the stage for functional studies.
Sigma-Aldrich
Kenpaullone, ≥98%
Sigma-Aldrich
ReNcell NSC Maintenance Media, ReNcell NSC Maintenance Medium is a defined serum-free, growth factor-free neural stem cell medium that has been optimized for the growth & in vitro differentiation of ReNcell immortalized human neural progenitor cells.