Merck
  • The circRNA interactome-innovative hallmarks of the intra- and extracellular radiation response.
The circRNA interactome-innovative hallmarks of the intra- and extracellular radiation response.
Oncotarget (2017-11-08)
Valerie Bríd O'Leary, Jan Smida, Martina Matjanovski, Corinna Brockhaus, Klaudia Winkler, Simone Moertl, Saak Victor Ovsepian, Michael John Atkinson
ABSTRACT
Generated by Quaking (QKI), circular RNAs (circRNAs) are newly recognised non-coding RNA (ncRNA) members characterised by tissue specificity, increased stability and enrichment within exosomes. Studies have shown that ionizing radiation (IR) can influence ncRNA transcription. However, it is unknown whether circRNAs or indeed QKI are regulated by IR. Microarray circRNA profiling and next generation sequencing revealed that circRNA expression was altered by low and medium dose exposure sourced predominantly from genes influencing the p53 pathway. CircRNAs KIRKOS-71 and KIRKOS-73 transcribed from the WWOX (WW Domain Containing Oxidoreductase) tumor suppressor (a p53 regulator) responded within hours to IR. KIRKOS-71 and KIRKOS-73 were present in exosomes yet exhibited differential transcript clearance between irradiated cell lines. Dual-quasar labelled probes and in-situ hybridization demonstrated the intercellular distribution of KIRKOS-71 and KIRKOS-73 predominantly within the perinucleus. QKI knockdown removed nuclear expression of these circRNAs with no significant effect on cytosolic KIRKOS-71 and KIRKOS-73. Distinct QKI transcription between cell lines and its augmented interaction with KIRKOS-71 and KIRKOS-73 was noted post IR. This foremost study provides evidence that QKI and circRNAs partake in the cellular irradiation response. KIRKOS-71 and KIRKOS-73 as stable secreted circRNAs may afford vital characteristics worth syphoning as promising diagnostic radiotherapy biomarkers.
Sigma-Aldrich
Anti-QKI-7 Antibody, serum, Chemicon®
Roche
cOmplete Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Magna RIP® RNA-Binding Protein Immunoprecipitation Kit, RNA Immunoprecipitation (RIP) Kit containing all necessary reagents to perform 12 individual RNA-binding protein immunoprecipitation (RIP) reactions using protein A/G magnetic beads.
Millipore
Nuclear Extraction Kit