Merck
  • Role of ATP-bound divalent metal ion in the conformation and function of actin. Comparison of Mg-ATP, Ca-ATP, and metal ion-free ATP-actin.
Role of ATP-bound divalent metal ion in the conformation and function of actin. Comparison of Mg-ATP, Ca-ATP, and metal ion-free ATP-actin.
The Journal of biological chemistry (1991-04-25)
C Valentin-Ranc, M F Carlier
ABSTRACT
The fluorescence of N-acetyl-N'-(sulfo-1-naphthyl)ethylenediamine (AEDANS) covalently bound to Cys-374 of actin is used as a probe for different conformational states of G-actin according to whether Ca-ATP, Mg-ATP, or unchelated ATP is bound to the nucleotide site. Upon addition of large amounts (greater than 10(2)-fold molar excess) of EDTA to G-actin, metal ion-free ATP-G-actin is obtained with EDTA bound. Metal ion free ATP-G-actin is characterized by a higher AEDANS fluorescence than Mg-ATP-G-actin, which itself has a higher fluorescence than Ca-ATP-G-actin. Evidence for EDTA binding to G-actin is shown using difference spectrophotometry. Upon binding of EDTA, the rate of dissociation of the divalent metal ion from G-actin is increased (2-fold for Ca2+, 10-fold for Mg2+) in a range of pH from 7.0 to 8.0. A model is proposed that quantitatively accounts for the kinetic data. The affinity of ATP is weakened 10(6)-fold upon removal of the metal ion. Metal ion-free ATP-G-actin is in a partially open conformation, as indicated by the greater accessibility of -SH residues, yet it retains functional properties of polymerization and ATP hydrolysis that appear almost identical to those of Ca-ATP-actin, therefore different from those of Mg-ATP-actin. These results are discussed in terms of the role of the ATP-bound metal ion in actin structure and function.
Sigma-Aldrich
Adenosine 5′-triphosphate di(tris) salt hydrate, ≥95%