Merck
  • Chemical-genetic disruption of clathrin function spares adaptor complex 3-dependent endosome vesicle biogenesis.
Chemical-genetic disruption of clathrin function spares adaptor complex 3-dependent endosome vesicle biogenesis.
Molecular biology of the cell (2013-06-14)
Stephanie A Zlatic, Emily J Grossniklaus, Pearl V Ryder, Gloria Salazar, Alexa L Mattheyses, Andrew A Peden, Victor Faundez
ABSTRACT
A role for clathrin in AP-3-dependent vesicle biogenesis has been inferred from biochemical interactions and colocalization between this adaptor and clathrin. The functionality of these molecular associations, however, is controversial. We comprehensively explore the role of clathrin in AP-3-dependent vesicle budding, using rapid chemical-genetic perturbation of clathrin function with a clathrin light chain-FKBP chimera oligomerizable by the drug AP20187. We find that AP-3 interacts and colocalizes with endogenous and recombinant FKBP chimeric clathrin polypeptides in PC12-cell endosomes. AP-3 displays, however, a divergent behavior from AP-1, AP-2, and clathrin chains. AP-3 cofractionates with clathrin-coated vesicle fractions isolated from PC12 cells even after clathrin function is acutely inhibited by AP20187. We predicted that AP20187 would inhibit AP-3 vesicle formation from endosomes after a brefeldin A block. AP-3 vesicle formation continued, however, after brefeldin A wash-out despite impairment of clathrin function by AP20187. These findings indicate that AP-3-clathrin association is dispensable for endosomal AP-3 vesicle budding and suggest that endosomal AP-3-clathrin interactions differ from those by which AP-1 and AP-2 adaptors productively engage clathrin in vesicle biogenesis.
Sigma-Aldrich
Anti-Synaptophysin Antibody, clone SY38, clone SY38, Chemicon®, from mouse
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Clathrin Light Chain Antibody, serum, Chemicon®
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Clathrin Heavy Chain Mouse mAb (X22), liquid, clone X22, Calbiochem®
Pricing and availability is not currently available.